Learn More
Hach Company Manifold, Filter Funnel, 316L Stainless Steel
Supplier:  Hach Company 2486100
Encompass
Feature 3-place filtration with independent vacuum control. Adapters are not included. (Adapters shown in the photo not included. These are elongated standard adapters).Accepts 25 or 47 mm filter funnels for processing multiple samples simultaneously. Designed for efficient analysis of microbiological or particulate contamination. Aluminum: Anodized aluminum body; chrome-plated brass valves; stainless steel drain plugs. Autoclavable. Dimensions (H x W x D) 3.9" x 17.2" x 6.0"
Catalog No. 50-199-6056