Learn More
Thomas Scientific MANIFOLD 384 WELL PLATE 24 Straight
SDP
Catalog No. NC0254043
Encompass
Manifolds for Drummond Microdispenser For use with 7691K20 Microdispenser and L55 Repeat Kit or L60 Pall Valve for multiple, simultaneous deliveries to microtest plates Manifolds are stainless steel, with removable Teflon end caps and Luer connector for attachment to pall valve Autoclavable Straight-type manifolds have a central intake shaft perpendicular to manifold and Teflon plug at either end of the manifold Bent-type manifolds have offset intake shaft, mounted at one end of the manifold and a Teflon plug at the other end This configuration minimizes contamination of sterile microtest plates Can be connected to vacuum line for use as an aspirator 7691N80 through N92 Manifolds have delivery spouts on 4.5 mm centers for use with 384-well plates. 7691R10 through R60 Manifolds have delivery spouts on 9 mm centers for use with 96-well plates. 7691R75 Manifold has delivery spouts on 20 mm centers for use with 24-well plates. Furnished with a cleaning wire which has attached finger
Catalog No. NC0254043 Supplier Thomas Scientific Supplier No. 7691N80
Manifolds for Drummond Microdispenser For use with 7691K20 Microdispenser and L55 Repeat Kit or L60 Pall Valve for multiple, simultaneous deliveries to microtest plates Manifolds are stainless steel, with removable Teflon end caps and Luer connector for attachment to pall valve Autoclavable Straight-type manifolds have a central intake shaft perpendicular to manifold and Teflon plug at either end of the manifold Bent-type manifolds have offset intake shaft, mounted at one end of the manifold and a Teflon plug at the other end This configuration minimizes contamination of sterile microtest plates Can be connected to vacuum line for use as an aspirator 7691N80 through N92 Manifolds have delivery spouts on 4.5 mm centers for use with 384-well plates. 7691R10 through R60 Manifolds have delivery spouts on 9 mm centers for use with 96-well plates. 7691R75 Manifold has delivery spouts on 20 mm centers for use with 24-well plates. Furnished with a cleaning wire which has attached finger