Learn More
Waters Corp Waters Extraction Plate Manifold, for Oasis 96-Well Plates
Catalog No. 50819115 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Extraction Plate Manifold is designed to support high-throughput solid-phase extraction (SPE) using Oasis sorbents.
  • Includes vacuum gauge, control, and release valves for monitoring and adjusting flow rates.
  • Three-point interlocking system ensures proper orientation of racks.
  • Compatible with vacuum pump 220V/240V 50Hz.
  • Package includes glass chamber with exit valve, white Delrin cover with gasket and valves, needle tips, luer plugs (male), and needle tip ejector tool.
  • A rack is packaged separately.
  • Designed for use with Oasis 96-Well Plates.
  • Includes a spacer for use with Oasis μElution Plates; spacer is removable for Oasis 96 Well Extraction Plates.
Catalog No. 50-819-115 Supplier Waters Corp Supplier No. 186001831
The Extraction Plate Manifold is designed to support high-throughput solid-phase extraction (SPE) using Oasis sorbents.
  • Includes vacuum gauge, control, and release valves for monitoring and adjusting flow rates.
  • Three-point interlocking system ensures proper orientation of racks.
  • Compatible with vacuum pump 220V/240V 50Hz.
  • Package includes glass chamber with exit valve, white Delrin cover with gasket and valves, needle tips, luer plugs (male), and needle tip ejector tool.
  • A rack is packaged separately.
  • Designed for use with Oasis 96-Well Plates.
  • Includes a spacer for use with Oasis μElution Plates; spacer is removable for Oasis 96 Well Extraction Plates.