Learn More
Braintree Scientific Manifold Assembly - 3 station unit
SDP
Catalog No. 501955019
Encompass
Manifold supports multiple procedures while utilizing only one vaporizer. While offering individual shut-offs, manifold provides intake of new gas to designated locations while opposite side provides output for used gases to a scavenger unit. (4) Patient Unit Shown **Sold for research purposes only and not for human use**
Catalog No. 50-195-5019 Supplier Braintree Scientific Supplier No. MAN3
Manifold supports multiple procedures while utilizing only one vaporizer. While offering individual shut-offs, manifold provides intake of new gas to designated locations while opposite side provides output for used gases to a scavenger unit. (4) Patient Unit Shown **Sold for research purposes only and not for human use**