Learn More
Chemglass Life Sciences 3-Port Vacuum Manifold, Double Bank, Airfree, Schlenk, 410mm OAL
SDP
Catalog No. AF030013
Encompass_Preferred
3-Port Vacuum Manifold, Double Bank, Airfree, Schlenk, 410mm OAL. Double bank 3-port manifold with 0-8mm Chem-Vac™ Chem-Cap valves for use on the AF-0300-02 portable cart. Valves face forward permitting convenient operation of both vacuum and inert gas lines. Requires the use of an AF-0300-14 adapter for connection to the AF-0300-04 trap assembly at the left top bank of the manifold. Ports have #15 o-ring joints and are spaced 100mm center to center while the manifold has an overall length of 410mm. Left side of manifold has #20 o-ring joints for the connection to AF-0300-14 adapters. Right side of manifold has #15 o-ring joints for use with the AF-0306 gauge adapter and AF-0210-01 stopper. Please note: Not supplied with O-Rings.
Catalog No. AF030013 Supplier Chemglass Life Sciences Supplier No. AF030013
3-Port Vacuum Manifold, Double Bank, Airfree, Schlenk, 410mm OAL. Double bank 3-port manifold with 0-8mm Chem-Vac™ Chem-Cap valves for use on the AF-0300-02 portable cart. Valves face forward permitting convenient operation of both vacuum and inert gas lines. Requires the use of an AF-0300-14 adapter for connection to the AF-0300-04 trap assembly at the left top bank of the manifold. Ports have #15 o-ring joints and are spaced 100mm center to center while the manifold has an overall length of 410mm. Left side of manifold has #20 o-ring joints for the connection to AF-0300-14 adapters. Right side of manifold has #15 o-ring joints for use with the AF-0306 gauge adapter and AF-0210-01 stopper. Please note: Not supplied with O-Rings.