Learn More
Ace Glass, Inc. Vacuum manifold with three variable opening, 0-10mm high vacuum threaded stopcock ports. 235mm overall length with left side hose connection. PTFE plugs with UHDPE handles.
SDP
Catalog No. 50976256
Encompass_Preferred
All-glass-and-PTFE vacuum manifold with three left side stopcock ports. Each port is a variable opening, 0-10mm, high vacuum threaded stopcock. Plugs are PTFE with Nylon or PTFE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with O-Rings as supplied.
Catalog No. 50-976-256 Supplier Ace Glass, Inc. Supplier No. 873012
All-glass-and-PTFE vacuum manifold with three left side stopcock ports. Each port is a variable opening, 0-10mm, high vacuum threaded stopcock. Plugs are PTFE with Nylon or PTFE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with O-Rings as supplied.