Learn More
Ace Glass, Inc. Manifold, air sampling, #25 Ace-Thred to 1inch tube end, 36inchlength, (7) #7 Ace-Thred ports
SDP
Catalog No. 50974860
Encompass_Preferred
Manifold, air sampling, #25 Ace-Thred to 1inch tube end, 36inchlength, (7) #7 Ace-Thred ports
Catalog No. 50-974-860 Supplier Ace Glass, Inc. Supplier No. 748814367
Manifold, air sampling, #25 Ace-Thred to 1inch tube end, 36inchlength, (7) #7 Ace-Thred ports