Learn More
Ace Glass, Inc. Manifold, air sampling, #25 Ace-Thred to 1inch tube end, 48inchlength, (3) #7 Ace-Thred ports
SDP
Catalog No. 50974863
Encompass_Preferred
Manifold, air sampling, #25 Ace-Thred to 1inch tube end, 48inchlength, (3) #7 Ace-Thred ports
Catalog No. 50-974-863 Supplier Ace Glass, Inc. Supplier No. 748814483
Manifold, air sampling, #25 Ace-Thred to 1inch tube end, 48inchlength, (3) #7 Ace-Thred ports