Learn More
Ace Glass, Inc. Manifold, air sampling, #25 Ace-Thred ends, 72inch length, (5)#7 Ace-Thred ports
SDP
Catalog No. 50979295
Encompass_Preferred
Manifold, air sampling, #25 Ace-Thred ends, 72inch length, (5)#7 Ace-Thred ports
Catalog No. 50-979-295 Supplier Ace Glass, Inc. Supplier No. 748834725
Manifold, air sampling, #25 Ace-Thred ends, 72inch length, (5)#7 Ace-Thred ports