Learn More
Ace Glass, Inc. Manifold, air sampling, #50 Ace-Thred to 2inch tube end, 24inchlength, (5) #7 Ace-Thred ports
SDP
Catalog No. 50974877
Encompass_Preferred
Manifold, air sampling, #50 Ace-Thred to 2inch tube end, 24inchlength, (5) #7 Ace-Thred ports
Catalog No. 50-974-877 Supplier Ace Glass, Inc. Supplier No. 748815245
Manifold, air sampling, #50 Ace-Thred to 2inch tube end, 24inchlength, (5) #7 Ace-Thred ports