Learn More
Ace Glass, Inc. Manifold, air sampling, #50 Ace-Thred to 2inch tube end, 48inchlength, (3) #7 Ace-Thred ports
SDP
Catalog No. 50974882
Encompass_Preferred
Manifold, air sampling, #50 Ace-Thred to 2inch tube end, 48inchlength, (3) #7 Ace-Thred ports
Catalog No. 50-974-882 Supplier Ace Glass, Inc. Supplier No. 748815483
Manifold, air sampling, #50 Ace-Thred to 2inch tube end, 48inchlength, (3) #7 Ace-Thred ports