Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polycarbonate, Five Ports, Manifolds with Female Luer Locks, 360 Rotation, Non-Sterile; 10/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3060043
Encompass
Polycarbonate manifolds have polyurethane stems and HDPE handles. Ports have female luer locks.
Catalog No. NC9755007