Learn More
Chemglass Life Sciences 3-Port Vacuum Manifold, Single Bank, Airfree, Schlenk
SDP
Supplier:  Chemglass Life Sciences AF030003
Encompass_Preferred
3-Port Vacuum Manifold, Single Bank, Airfree, Schlenk. Single bank 3-port manifold with 0-8mm Chem-Vac™ Chem-Cap valves as ports. A 0-20mm shut off valve is on the left side of the manifold for connection to the AF-0300-04 vacuum trap via a #20 o-ring joint. Ports all have #15 o-ring joints and can be used with AF-0070 through AF-0080 adapters for connection to standard taper joints, serrated hose connections, etc. Spacing on the ports is 6" center to center while the overall length of the manifold is 470mm. Right side of the manifold has a #15 o-ring joint for connection to a AF-0306-01 thermocouple gauge adapter.PLEASE NOTE: NOT SUPPLIED WITH O-RINGS.
Catalog No. 50-153-0947