Learn More
Synthware™ Manifold with Solid Glass Stopcocks
GREENER_CHOICESDP
Catalog No. 31501804 Shop All Kemtech Products
Change view
Click to view available options
Material:
Borosilicate Glass
Catalog No. Material
31-501-804 Borosilicate Glass
1 options
Catalog No. 31-501-804 Supplier Synthware™ Supplier No. M150004
Add to Cart
Add to Cart
Features 4 mm double hollow high vacuum stopcocks.
Four–port manifold is fitted with two 14/20 and 24/40 inner joints, and is 100 mm center to center joints.
Specifications
Description Front Left and Right; Rear Left and Right Spherical Connection
Type Vacuum/Gas Manifold Double, O-ring
For Use With (Application) Organic Synthesis
Includes 4 ports
Material Borosilicate Glass