Learn More
Chemglass Life Sciences High Flow Manifold System with Quick Connect Fittings, 10L-20L Reactor Capacity, M16 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200°C Temperature Range
SDP
Catalog No. 501531350
Encompass_Preferred
High Flow Manifold System with Quick Connect Fittings, 10L-20L Reactor Capacity, M16 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200C Temperature Range. The Chemglass high flow manifold system allows for easy and safe connection of your circulator to Chemglass's glass process reactors. Manifold helps alleviate excessive mechanical forces placed on the glass vessel inlet/outlet. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on Stainless Steel Beaded Pipe fitting on one end and Stainless Steel 3/4 Male NPT fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately. CG-1969-M-10 Fits Support Frame CG-1965 and CG-1965-X.
Catalog No. 50-153-1350 Supplier Chemglass Life Sciences Supplier No. CG1969M10
High Flow Manifold System with Quick Connect Fittings, 10L-20L Reactor Capacity, M16 Circulator Size End Fitting, 1" Beaded Pipe Triple Insulated Flexible Hose, -60 to 200C Temperature Range. The Chemglass high flow manifold system allows for easy and safe connection of your circulator to Chemglass's glass process reactors. Manifold helps alleviate excessive mechanical forces placed on the glass vessel inlet/outlet. System is supplied complete as illustrated with Triple Insulated Flexible Hose. Hoses have welded on Stainless Steel Beaded Pipe fitting on one end and Stainless Steel 3/4 Male NPT fitting on the other. Hoses to connect manifold to circulator are NOT supplied and must be ordered separately. CG-1969-M-10 Fits Support Frame CG-1965 and CG-1965-X.