Learn More
Chemglass Life Sciences Manifold System, Benchtop, 300mL-5L Reactor Capacity, M16 Circulator Size End Fitting, -60 to 200°C Temperature Range
SDP
Catalog No. 501531351
Encompass_Preferred
Manifold System, Benchtop, 300mL-5L Reactor Capacity, M16 Circulator Size End Fitting, -60 to 200C Temperature Range. The Chemglass manifold system allows for easy and safe connection of your circulator to Chemglass's glass benchtop reactors. Manifold provides a convenient way to monitor jacket temperature and pressure. Triple Insulated Flexible Hose for connecting manifold to circulator and reactor are sold separately. Manifold assembly mounts into Chemglass's CG-9253 Kwik Klamp II or CG-9252 clamps. To allow flexibility for different reactor configurations the inlet and outlet must be installed by the end user, the list below shows the list of connections supplied. Thread sealant is supplied with manifold block to ensure leak free assembly. PLEASE NOTE: TEMPERATURE LIMITATIONS ON MANIFOLD IS -60 TO 200C.
Catalog No. 50-153-1351 Supplier Chemglass Life Sciences Supplier No. CG1969M14
Manifold System, Benchtop, 300mL-5L Reactor Capacity, M16 Circulator Size End Fitting, -60 to 200C Temperature Range. The Chemglass manifold system allows for easy and safe connection of your circulator to Chemglass's glass benchtop reactors. Manifold provides a convenient way to monitor jacket temperature and pressure. Triple Insulated Flexible Hose for connecting manifold to circulator and reactor are sold separately. Manifold assembly mounts into Chemglass's CG-9253 Kwik Klamp II or CG-9252 clamps. To allow flexibility for different reactor configurations the inlet and outlet must be installed by the end user, the list below shows the list of connections supplied. Thread sealant is supplied with manifold block to ensure leak free assembly. PLEASE NOTE: TEMPERATURE LIMITATIONS ON MANIFOLD IS -60 TO 200C.