Learn More
Ace Glass, Inc. Bakeable vacuum manifold with five variable opening, 0-10mmhigh vacuum threaded stopcock ports. 375mm overall length with right side hose connection. PTFE plugs with PTFE handles.
SDP
Catalog No. 50976264
Encompass_Preferred
All-glass-and-PTFE vacuum bakeable manifold with five right side stopcock ports. Each port is a variable opening, 0-10mm, high vacuum threaded stopcock. Plugs are PTFE with Nylon or PTFE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with O-Rings as supplied.
Catalog No. 50-976-264 Supplier Ace Glass, Inc. Supplier No. 873037
All-glass-and-PTFE vacuum bakeable manifold with five right side stopcock ports. Each port is a variable opening, 0-10mm, high vacuum threaded stopcock. Plugs are PTFE with Nylon or PTFE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with O-Rings as supplied.