Learn More
Ace Glass, Inc. Bakeable vacuum manifold with three variable opening, 0-10mmhigh vacuum threaded stopcock ports. 235mm overall length with left side hose connection. PTFE plugs with PTFE handles.
SDP
Catalog No. 50979825
Encompass_Preferred
All-glass-and-PTFE vacuum bakeable manifold with three left side stopcock ports. Each port is a variable opening, 0-10mm, high vacuum threaded stopcock. Plugs are PTFE with Nylon or PTFE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with O-Rings as supplied.
Catalog No. 50-979-825 Supplier Ace Glass, Inc. Supplier No. 873014
All-glass-and-PTFE vacuum bakeable manifold with three left side stopcock ports. Each port is a variable opening, 0-10mm, high vacuum threaded stopcock. Plugs are PTFE with Nylon or PTFE handles and FETFE O-Rings. Ultimate vacuum of 10-7 can be realized with O-Rings as supplied.