Learn More
Chemglass Life Sciences 4-Port Vacuum Manifold, Right Handed Trap Assembly, Double Bank, Airfree, Schlenk, Complete
SDP
Supplier:  Chemglass Life Sciences AF045401
Encompass_Preferred
4-Port Vacuum Manifold, Right Handed Trap Assembly, Double Bank, Airfree, Schlenk, Complete, Manifold completye with the dual trap assrmbly is 46" W x 23" H. Supplied completwe with two # 18" stainless stell pinch clamps,three # 35" stainless stell pinch clamps, and one 65" S.S.pinch clamps, two # 112 Viyon o-rings,and one #226 Viton o-ring.
Catalog No. 50-872-175