Learn More
Chemglass Life Sciences 5-Port Vacuum Manifold, Right Handed Trap Assembly, Double Bank, Airfree, Schlenk
SDP
Catalog No. 501530956
Encompass_Preferred
Line has 5-ports w/4mm high vacuum stopcocks having a 24/40 inner joints w/ hooks just above grind. Ports are spaced 150mm center to center & have rod bracing to vacuum bank. 40mm ID vacuum bank evacuates vessels up to 1000ml. End of vacuum bank has 35/25 ball joint & supplied w/socket joint cap style stopper. Valved gauging port is placed on main vacuum line & has a 12/5 ball joint. Inert gas line has 2-ports for connection of inert gas source & bubbler. End of inert gas line has a 14/20 inner joint & is supplied w/a joint cap style stopper. Pre-trap is 51mm OD x 250mm long w/ two 28/15 socket joints. Main trap body is 58 mm OD x 235mm long w/a reversed 75/50 ball & socket joint connection. A valved gauging port is placed between main & pre-trap. Spacing between the two traps is 175mm for easy placement of dewar flasks. OAD: 40" W x 24" H. Also supplied w/ two #28 S.S. pinch clamps, one #35 S.S. pinch clamp, one #75 S.S. pinch clamp, & two 1/2" S.S. springs.
Catalog No. 50-153-0956 Supplier Chemglass Life Sciences Supplier No. AF045001
Add to Cart
Add to Cart
Request a Quote
Line has 5-ports w/4mm high vacuum stopcocks having a 24/40 inner joints w/ hooks just above grind. Ports are spaced 150mm center to center & have rod bracing to vacuum bank. 40mm ID vacuum bank evacuates vessels up to 1000ml. End of vacuum bank has 35/25 ball joint & supplied w/socket joint cap style stopper. Valved gauging port is placed on main vacuum line & has a 12/5 ball joint. Inert gas line has 2-ports for connection of inert gas source & bubbler. End of inert gas line has a 14/20 inner joint & is supplied w/a joint cap style stopper. Pre-trap is 51mm OD x 250mm long w/ two 28/15 socket joints. Main trap body is 58 mm OD x 235mm long w/a reversed 75/50 ball & socket joint connection. A valved gauging port is placed between main & pre-trap. Spacing between the two traps is 175mm for easy placement of dewar flasks. OAD: 40" W x 24" H. Also supplied w/ two #28 S.S. pinch clamps, one #35 S.S. pinch clamp, one #75 S.S. pinch clamp, & two 1/2" S.S. springs.