Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALADDIN SCIENTIFIC CORPORATION Mannitol, 100g

Catalog No. 5000082575
Encompass_Preferred

Mannitol Ultra pure 99%(HPLC)Cas-69-65-8

Catalog No. 50-000-82575 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. M108829100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mannitol Ultra pure 99%(HPLC)Cas-69-65-8

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.