Learn More
Ops Diagnostics MANNITOL 500G USP GRAD LYOPHI
Catalog No. NC2402358
Encompass
Mannitol USP Grade and Lyophilization Certified is a sugar alcohol (polyol) that is extremely useful for freeze drying proteins and other biomolecules. It is widely used as an excipient in pharmaceutical applications because it allows proteins peptides and other biomolecules to be preserved without succumbing to chemical and physical degradation over time.
Catalog No. NC2402358 Supplier Ops Diagnostics Supplier No. EXMN 50001
Mannitol USP Grade and Lyophilization Certified is a sugar alcohol (polyol) that is extremely useful for freeze drying proteins and other biomolecules. It is widely used as an excipient in pharmaceutical applications because it allows proteins peptides and other biomolecules to be preserved without succumbing to chemical and physical degradation over time.