Learn More
NEOGEN Mannitol Salt Agar, 10 kg
SDP
Catalog No. 502015383
Encompass_Preferred
Mannitol Salt Agar is used for the isolation of staphylococci in a laboratory setting. Mannitol Salt Agar is not intended for use in the diagnosis of disease or other conditions in humans. Conforms to Harmonized USP/EP/JP Requirements.Chapman formulated Mannitol Salt Agar to isolate staphylococci by inhibiting growth of most other bacteria with a high salt concentration. Chapman added 7.5% Sodium Chloride to Phenol Red Mannitol Agar and noted pathogenic strains of staphylococci (coagulase-positive staphylococci) grew luxuriantly and produced yellow colonies with yellow zones. Non-pathogenic staphylococci produced small red colonies with no color change to the surrounding medium. Mannitol Salt Agar is recommended for isolating pathogenic staphylococci from samples, cosmetics, and microbial limit tests.For more information visit https://www.neogen.com/solutions/Microbiology/mannitol-salt-agar
Catalog No. 50-201-5383 Supplier NEOGEN Supplier No. 700003187
Mannitol Salt Agar is used for the isolation of staphylococci in a laboratory setting. Mannitol Salt Agar is not intended for use in the diagnosis of disease or other conditions in humans. Conforms to Harmonized USP/EP/JP Requirements.Chapman formulated Mannitol Salt Agar to isolate staphylococci by inhibiting growth of most other bacteria with a high salt concentration. Chapman added 7.5% Sodium Chloride to Phenol Red Mannitol Agar and noted pathogenic strains of staphylococci (coagulase-positive staphylococci) grew luxuriantly and produced yellow colonies with yellow zones. Non-pathogenic staphylococci produced small red colonies with no color change to the surrounding medium. Mannitol Salt Agar is recommended for isolating pathogenic staphylococci from samples, cosmetics, and microbial limit tests.For more information visit https://www.neogen.com/solutions/Microbiology/mannitol-salt-agar