Learn More
Bioworld Mannitol Salt Agar, 500 g
Catalog No. 50254333
Encompass_Preferred
Mannitol Salt AgarSelective agar medium used for the identification and isolation of pathogenic staphylococcipH: 7.4Working Concentration: 111 g/LPrepared Appearance: pinkish red agarSoluble in boiling waterComposition (g/L):Pancreatic Digest of Casei: 5g Peptic Digest of Animal Tissue: 5g Beef Extract: 1g D-Mannitol: 10gSodium Chloride: 75gPhenol Red: 0.025gAgar: 15g
Catalog No. 50-254-333 Supplier Bioworld Supplier No. 306200511
Mannitol Salt AgarSelective agar medium used for the identification and isolation of pathogenic staphylococcipH: 7.4Working Concentration: 111 g/LPrepared Appearance: pinkish red agarSoluble in boiling waterComposition (g/L):Pancreatic Digest of Casei: 5g Peptic Digest of Animal Tissue: 5g Beef Extract: 1g D-Mannitol: 10gSodium Chloride: 75gPhenol Red: 0.025gAgar: 15g