Learn More
Andwin Scientific MANNITOL SALT AGAR 2 KG
SDP
Catalog No. NC0733868
Encompass_Preferred
Mannitol Salt Agar is a medium made to isolate and differentiate both pathogenic and non-pathogenic Staphylococci. On this medium, Staphylococcus aureus strains product large yellow colonies surrounded by yellow zones while non-pathogenic Staphylococci product smaller, red colonies. This product is sold in a quantity of 2 kilograms.
Catalog No. NC0733868 Supplier Andwin Scientific Supplier No. M13114
Mannitol Salt Agar is a medium made to isolate and differentiate both pathogenic and non-pathogenic Staphylococci. On this medium, Staphylococcus aureus strains product large yellow colonies surrounded by yellow zones while non-pathogenic Staphylococci product smaller, red colonies. This product is sold in a quantity of 2 kilograms.