Learn More
MOLTOX Molecular Toxicology Inc Mannitol Salt Agar Plate, 20 mL/plate, 20 plates/sleeve, Store at Room Temperature
SDP
Catalog No. 50948680
Encompass_Preferred
Mannitol Salt Agar is a selective isolation and an enumerating agar for Staphylococcus. It also aids in the differentiation of staphylococcal species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-680 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S64.141
Mannitol Salt Agar is a selective isolation and an enumerating agar for Staphylococcus. It also aids in the differentiation of staphylococcal species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.