Learn More
Teknova Mannitol Salt Agar Plates. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387286
Encompass
1.0% D-Mannitol, 1.0% Proteose Peptone, 0.1% Beef Extract, 7.5% Sodium Chloride, 25g/mL Phenol Red, 1.5% Agar
Catalog No. 50-238-7286 Supplier Teknova Supplier No. M59204PK
1.0% D-Mannitol, 1.0% Proteose Peptone, 0.1% Beef Extract, 7.5% Sodium Chloride, 25g/mL Phenol Red, 1.5% Agar