Learn More
Teknova Mannitol Soy Agar Plates with 10mM Magnesium Chloride. 100mm, 30mL fill, 20 Plates/Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387758
Encompass
2.0% Mannitol, 2.0% Soy Flour,2.0% Agar, 10mM Magnesium Chloride
Catalog No. 50-238-7758 Supplier Teknova Supplier No. M10354PK
2.0% Mannitol, 2.0% Soy Flour,2.0% Agar, 10mM Magnesium Chloride