Learn More
U.S. Pharmacopeia Mannitol, 69-65-8, MFCD00064287, 1.5g
Catalog No. NC9083436
Encompass
Molecular formula C6H14O6, Molecular weight 182.2, Melting Point 330.8 - 338 °F (166 - 170 °C), Boiling Point 554 - 563 °F (290 - 295 °C) 0.466628 kPa, Synonyms: 1,2,3,4,5,6-Hexanehexol, Mannite
Catalog No. NC9083436 Supplier U.S. Pharmacopeia Supplier No. 1375105
Molecular formula C6H14O6, Molecular weight 182.2, Melting Point 330.8 - 338 °F (166 - 170 °C), Boiling Point 554 - 563 °F (290 - 295 °C) 0.466628 kPa, Synonyms: 1,2,3,4,5,6-Hexanehexol, Mannite