Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-Mannosidase | 9025-42-7 | 1 MG
SDP

Catalog No. 5000400129
Encompass_Preferred

α-Mannosidase is a hydrolytic enzyme that targets mannose-containing glycoproteins or glycolipids. It is considered promising for research related to inherited α-mannosidosis.

  • Hydrolytic enzyme
  • Targets mannose-containing glycoproteins or glycolipids
  • Promising for research of inherited α-mannosidosis
  • Source: Jack bean
  • Specific activity: 18 unit/mg protein
  • Appearance: Colorless to light yellow liquid
  • For research use only

Catalog No. 50-004-00129 Supplier Medchemexpress LLC Supplier No. HYP29501MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Mannosidase is a hydrolytic enzyme that targets mannose-containing glycoproteins or glycolipids. It is considered promising for research related to inherited α-mannosidosis.

  • Hydrolytic enzyme
  • Targets mannose-containing glycoproteins or glycolipids
  • Promising for research of inherited α-mannosidosis
  • Source: Jack bean
  • Specific activity: 18 unit/mg protein
  • Appearance: Colorless to light yellow liquid
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.