Learn More
Electron Microscopy Sciences Manomark™ Pen Assorted
SDP
Catalog No. 50949520
Encompass_Preferred
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins. With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it's left uncapped. Once dry, the solvent base ink is temperature resistant from 0 to 155 degree C (32-310 degree F). Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.
Catalog No. 50-949-520 Supplier Electron Microscopy Sciences Supplier No. 72127AS
Writes on all plastic, glass, metal, wool, paper and cloth, including "PTFE", fluoropolymer resins. With its precision seal valve the pens life is preserved and as well it allows for a smooth flow of ink even if it's left uncapped. Once dry, the solvent base ink is temperature resistant from 0 to 155 degree C (32-310 degree F). Markings are insoluble in water or hydrocarbons, resist many solvents, and are easily removed with alcohol.