Learn More
Medchemexpress LLC Manool | 596-85-0 | 290.48 | 50 MG
SDP
Catalog No. 5000416183
Encompass_Preferred
Manool is a diterpene derived from Salvia officinalis. It selectively induces cytotoxicity in cancer cells and arrests them at the G(2)/M phase of the cell cycle. Manool has shown higher cytotoxic activity against HeLa (IC50=6.7 μg/mL) and U343 (IC50=6.7 μg/mL) cells. Additionally, it exhibits a protective effect against chromosome damage induced by MMS in HepG2 cells.
  • Diterpene from Salvia officinalis.
  • Induces selective cytotoxicity in cancer cells.
  • Arrests cancer cells at the G(2)/M phase of the cell cycle.
  • Exhibits protective effect against chromosome damage.
Catalog No. 50-004-16183 Supplier Medchemexpress LLC Supplier No. HYN1039A50MG
Manool is a diterpene derived from Salvia officinalis. It selectively induces cytotoxicity in cancer cells and arrests them at the G(2)/M phase of the cell cycle. Manool has shown higher cytotoxic activity against HeLa (IC50=6.7 μg/mL) and U343 (IC50=6.7 μg/mL) cells. Additionally, it exhibits a protective effect against chromosome damage induced by MMS in HepG2 cells.
  • Diterpene from Salvia officinalis.
  • Induces selective cytotoxicity in cancer cells.
  • Arrests cancer cells at the G(2)/M phase of the cell cycle.
  • Exhibits protective effect against chromosome damage.