Learn More
Aviva Systems Biology Corp MANSC1 Antibody - N-terminal region (ARP34359_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34359_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MANSC1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MANSC1. Alternate names: LOH12CR3, 9130403P13RikApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52951