Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formulatrix MANTIS CHIP PFE LV .1 .5UL
SDP

Catalog No. NC2083693
Encompass

  • Diaphragm material: Perfluoroelastomer (PFE)
  • Volume range: 0.1 µL, 0.5 µL
  • Chemical compatibility: Maximum for harsh reagents
  • RFID tag: Included
  • Pallet integration: true
  • Dispensing speed: 10 dispenses/second
  • Dead volume: 6 µL with pipette tips
  • Accuracy error at 0.1 µL: 5%
  • Accuracy error at 0.5 µL: 4%
  • Coefficient of variation: <5%
  • Related chip lifetime: 750,000 cycles
  • Fluid channel diameter: 0.25 mm
  • Chip body material: Polypropylene plastic
  • Packaging: Pack of 6

Catalog No. NC2083693 Supplier Formulatrix Supplier No. 233129
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Diaphragm material: Perfluoroelastomer (PFE)
  • Volume range: 0.1 µL, 0.5 µL
  • Chemical compatibility: Maximum for harsh reagents
  • RFID tag: Included
  • Pallet integration: true
  • Dispensing speed: 10 dispenses/second
  • Dead volume: 6 µL with pipette tips
  • Accuracy error at 0.1 µL: 5%
  • Accuracy error at 0.5 µL: 4%
  • Coefficient of variation: <5%
  • Related chip lifetime: 750,000 cycles
  • Fluid channel diameter: 0.25 mm
  • Chip body material: Polypropylene plastic
  • Packaging: Pack of 6

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.