Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Formulatrix MANTIS CHIP-SILICONE LV
SDP

Catalog No. NC1914955
Encompass

  • Chip type: Silicone LV
  • Material: Silicone diaphragm
  • Dispense volume range: 0.1 uL, 0.5 uL
  • Grade: Molecular grade
  • Contamination testing: Free of DNA, DNase, RNA, RNase, and protease
  • Packaging: Sterile
  • Included components: Pre-attached molecular grade input tubing
  • Quantity per pack: 6 chips, 6 tubing pieces
  • RFID tag: Yes
  • Dispensing technology: Microfluidic valve cluster with micro-diaphragms
  • Maximum dispensing speed: 10 times per second
  • Compatibility: MANTIS instruments with RFID
  • Application focus: Assays requiring contamination-free liquid handling

Catalog No. NC1914955 Supplier Formulatrix Supplier No. 233649
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Chip type: Silicone LV
  • Material: Silicone diaphragm
  • Dispense volume range: 0.1 uL, 0.5 uL
  • Grade: Molecular grade
  • Contamination testing: Free of DNA, DNase, RNA, RNase, and protease
  • Packaging: Sterile
  • Included components: Pre-attached molecular grade input tubing
  • Quantity per pack: 6 chips, 6 tubing pieces
  • RFID tag: Yes
  • Dispensing technology: Microfluidic valve cluster with micro-diaphragms
  • Maximum dispensing speed: 10 times per second
  • Compatibility: MANTIS instruments with RFID
  • Application focus: Assays requiring contamination-free liquid handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.