Learn More
Formulatrix MANTIS HV CHIP TUBING 10
SDP
Catalog No. NC0918895
Encompass
  • Input tubing designed for precise fluidic delivery
  • Compatible with MANTIS HV diaphragm chips
  • Green color for easy identification
  • Inner diameter of 0.030 in
  • Length of 10 foot, with custom cut capability
  • Ideal for high-volume microfluidic applications
  • Versatile for various experimental configurations
Catalog No. NC0918895 Supplier Formulatrix Supplier No. 808978A
  • Input tubing designed for precise fluidic delivery
  • Compatible with MANTIS HV diaphragm chips
  • Green color for easy identification
  • Inner diameter of 0.030 in
  • Length of 10 foot, with custom cut capability
  • Ideal for high-volume microfluidic applications
  • Versatile for various experimental configurations