Learn More
Formulatrix MANTIS INPUT TUBES HV 6/PK
SDP
Supplier:  Formulatrix 232509
Encompass
  • Product type: Input tubing
  • System compatibility: MANTIS
  • Chip compatibility: HV diaphragm chips
  • Length: 45 cm
  • Quantity: 6 per pack
  • Application: High-volume liquid handling
  • Reservoir compatibility: Conical tubes
  • Manufacturer: FORMULATRIX
Catalog No. NC1889472