Learn More
Formulatrix MANTIS NOZZLE KIT, SILICONE
SDP
Catalog No. NC1971683
Encompass
Mantis Nozzle Kit, Silicone (Includes 0.1, 0.2, 0.3, 0.4, 0.6mm ID, 2 each)
Catalog No. NC1971683 Supplier Formulatrix Supplier No. 233036
Add to Cart
Add to Cart
Mantis Nozzle Kit, Silicone (Includes 0.1, 0.2, 0.3, 0.4, 0.6mm ID, 2 each)