Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100D RX50012
Encompass_Preferred
3 place multi position combo mantle for 500ml flat bottom flask, to 400 degrees C internal operating temperature, 180W 120V
Catalog No. NC1787091