Learn More
Ace Glass, Inc. Mantle support for Glas-Col M series mantles, fits Ace Glass12043-05, 12043-07 and Glas-Col 100C M98, 100C M100
SDP
Catalog No. 50977421
Encompass
Mantle support for Glas-Col M series mantles, fits Ace Glass12043-05, 12043-07 and Glas-Col 100C M98, 100C M100
Catalog No. 50-977-421 Supplier Ace Glass, Inc. Supplier No. 1209402
Mantle support for Glas-Col M series mantles, fits Ace Glass12043-05, 12043-07 and Glas-Col 100C M98, 100C M100