Learn More
Labsmith Tools For Science Manual 3-Port Selector Valve
SDP
Catalog No. NC1545515
Encompass
3-port manual valve with "L" flow pattern. Available for use with 360um capillary, 1/32" tubing, or 1/16" tubing via one-piece fittings (not included). Can be mounted to LabSmith breadboards.
Catalog No. NC1545515 Supplier Labsmith Tools For Science Supplier No. MV201C360
3-port manual valve with "L" flow pattern. Available for use with 360um capillary, 1/32" tubing, or 1/16" tubing via one-piece fittings (not included). Can be mounted to LabSmith breadboards.