Learn More
Azzota Corp Manual Abbe Refractometer, Measuring System: Optical Refraction Critical Angle Detection, Measuring Range: 1.3000-1.7000 Nd; 0.0-95% Brix, Resolution: 0.0005 Nd; 0.25% Brix; Accuracy: 0.0002 Nd; 0.10%
Catalog No. 507532968
Encompass_Preferred
Manual Abbe Refractometer, Measuring System: Optical refraction critical angle detection, Measuring Range: 1.3000-1.7000 nD; 0.0-95% Brix, Resolution: 0.0005 nD; 0.25% Brix; Accuracy: 0.0002 nD; 0.10%
Catalog No. 50-753-2968 Supplier Azzota Corp Supplier No. AR1
Manual Abbe Refractometer, Measuring System: Optical refraction critical angle detection, Measuring Range: 1.3000-1.7000 nD; 0.0-95% Brix, Resolution: 0.0005 nD; 0.25% Brix; Accuracy: 0.0002 nD; 0.10%