Learn More
Grainger Manual Air Control 1/8
Catalog No. 502751016
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve is a 1/4" NPT manual valve that controls air flow. It has a nickel-plated brass body with a chrome-plated brass ball, PTFE seals, and a knurled polymer handle. The valve has a Cv of 0.3 and a maximum pressure rating of 250 psi. It can be used to manually adjust and control air flow in pneumatic systems. The compact size makes it suitable for applications with space constraints.
Catalog No. 50-275-1016 Supplier Grainger Supplier No. 14F304
The Pneumadyne Inc. Manual Air Control Valve is a 1/4" NPT manual valve that controls air flow. It has a nickel-plated brass body with a chrome-plated brass ball, PTFE seals, and a knurled polymer handle. The valve has a Cv of 0.3 and a maximum pressure rating of 250 psi. It can be used to manually adjust and control air flow in pneumatic systems. The compact size makes it suitable for applications with space constraints.