Learn More
Grainger Manual Air Control 3-Way 2-Position 1/8
Catalog No. 502750726
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve is a 3-way valve that allows manual control of air flow. It has brass construction with a chrome-plated finish and includes a locking handle to hold valve position. This valve has 1/4" NPT female pipe thread ports and a maximum pressure rating of 250 PSI. The operating temperature range is -20 to 180°F. Key features include the ability to manually adjust air flow, a locking handle for holding valve position, brass and chrome construction, and 1/4" NPT ports. This valve provides manual air flow control for industrial pneumatic applications.
Catalog No. 50-275-0726 Supplier Grainger Supplier No. 14F275
The Pneumadyne Inc. Manual Air Control Valve is a 3-way valve that allows manual control of air flow. It has brass construction with a chrome-plated finish and includes a locking handle to hold valve position. This valve has 1/4" NPT female pipe thread ports and a maximum pressure rating of 250 PSI. The operating temperature range is -20 to 180°F. Key features include the ability to manually adjust air flow, a locking handle for holding valve position, brass and chrome construction, and 1/4" NPT ports. This valve provides manual air flow control for industrial pneumatic applications.