Learn More
Grainger Manual Air Control 3-Way 2-Position 1/8
Catalog No. 502750786
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve Three-Way is a manually operated pneumatic valve that controls the direction of airflow in pneumatic systems. It has a brass body construction with a chrome-plated brass ball, PTFE seals, and 1/4" NPT female ports. The valve allows switching between two output ports by rotating the lever handle 90 degrees. It has a Cv of 0.3 and a maximum pressure rating of 150 PSI. The valve can be used for a variety of directional air control applications.
Catalog No. 50-275-0786 Supplier Grainger Supplier No. 14F268
The Pneumadyne Inc. Manual Air Control Valve Three-Way is a manually operated pneumatic valve that controls the direction of airflow in pneumatic systems. It has a brass body construction with a chrome-plated brass ball, PTFE seals, and 1/4" NPT female ports. The valve allows switching between two output ports by rotating the lever handle 90 degrees. It has a Cv of 0.3 and a maximum pressure rating of 150 PSI. The valve can be used for a variety of directional air control applications.