Learn More
Grainger Manual AirControl 3-Way 2-Position 10-32
Catalog No. 502750997
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve 300 is a manually operated pneumatic valve designed for general purpose flow control applications. It has a brass body construction with a maximum pressure rating of 150 PSI. The valve features a knurled adjustment knob for precise flow control, 1/4" NPT female ports, and a flow coefficient (Cv) of 0.3. This compact valve can be used to control air flow in pneumatic systems. With its durable construction and precise adjustability, the Pneumadyne Manual Air Control Valve 300 provides reliable flow control for industrial automation applications.
Catalog No. 50-275-0997 Supplier Grainger Supplier No. 14F273
The Pneumadyne Inc. Manual Air Control Valve 300 is a manually operated pneumatic valve designed for general purpose flow control applications. It has a brass body construction with a maximum pressure rating of 150 PSI. The valve features a knurled adjustment knob for precise flow control, 1/4" NPT female ports, and a flow coefficient (Cv) of 0.3. This compact valve can be used to control air flow in pneumatic systems. With its durable construction and precise adjustability, the Pneumadyne Manual Air Control Valve 300 provides reliable flow control for industrial automation applications.