Learn More
Grainger Manual AirControl 3-Way 2-Position 10-32
Catalog No. 502749184
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve 0-150 PSI is a manually operated pneumatic valve that controls air pressure. It has a 0-150 PSI pressure range with 1/8" NPT female ports. The valve body and knob are made from aluminum and brass for durability. This valve allows precise air pressure regulation through manual adjustment of the knob. It can be panel mounted and is suitable for use in industrial automation applications that require manual air flow control. The compact size saves space.
Catalog No. 50-274-9184 Supplier Grainger Supplier No. 14F267
The Pneumadyne Inc. Manual Air Control Valve 0-150 PSI is a manually operated pneumatic valve that controls air pressure. It has a 0-150 PSI pressure range with 1/8" NPT female ports. The valve body and knob are made from aluminum and brass for durability. This valve allows precise air pressure regulation through manual adjustment of the knob. It can be panel mounted and is suitable for use in industrial automation applications that require manual air flow control. The compact size saves space.