Learn More
Grainger Manual AirControl 5-Way 2-Position 10-32
Catalog No. 502750844
Encompass_Preferred
The Pneumadyne Inc. Manual Air Control Valve is a manually operated valve that controls air flow. It has a brass body with a chrome-plated steel handle, 1/4" NPT female inlet and outlet ports, and a maximum pressure rating of 250 PSI. This valve allows you to adjust air flow by turning the handle to open and close the valve. It can be used for a variety of industrial and commercial air control applications. The valve body measures 2-1/2" long x 1-1/2" wide x 2" high.
Catalog No. 50-275-0844 Supplier Grainger Supplier No. 14F271
The Pneumadyne Inc. Manual Air Control Valve is a manually operated valve that controls air flow. It has a brass body with a chrome-plated steel handle, 1/4" NPT female inlet and outlet ports, and a maximum pressure rating of 250 PSI. This valve allows you to adjust air flow by turning the handle to open and close the valve. It can be used for a variety of industrial and commercial air control applications. The valve body measures 2-1/2" long x 1-1/2" wide x 2" high.