Learn More
MilliporeSigma™ Supelco™ Manual Bubble Flowmeter, Capillary Version
Catalog No. 111004266 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Includes:
Flowmeter, Stand
Catalog No. Includes
11-100-4266 Flowmeter, Stand
1 options
Catalog No. 11-100-4266 Supplier MilliporeSigma™ Supelco™ Supplier No. 23771
Add to Cart
Add to Cart
Determines volumetric flow rates by measuring the time taken by the gas stream to move a soap bubble through a specific volume
  • A simple and precise way to measure flow rates of gases
  • Allows the true split ratio to be determined by measuring actual flow rate
  • The flow tube is graduated in 0.01 mL increments to accomodate low flows that are typical of capillary GC applications
  • Two magnetic clamps hold the flow tube together so that the unit can be attached to any metal surface for easy storage
  • A version that includes a sampling stand is offered for added portability
TRUSTED_SUSTAINABILITY
Specifications
Type Bubble Flowmeter
For Use With (Application) Measuring Gas Flow Rates
Tube Size 0.5 mL
Includes Flowmeter, Stand