Learn More
Thomas Scientific Propette Manual Controller
SDP
Catalog No. 501457690
Encompass
ProPette™ MPC is the precise and easy to use manual pipette controller that 's always ready to use. After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting – there are no wires, no tubing and no battery to recharge! Begin aspirating by touching the EZ-Flow™ lever which is designed to precisely control the filling speed, based on how gently or firmly you press the lever. A built-in replaceable filter ensures optimum flow rate and protects the controller from liquid penetration and over-fill. A separate button provides a positive blow-out function when required. Made to accept pipettes from 0.1 to 100ml, the silicone "chuck" adapter is flexible, completely autoclavable, and will accommodate all types of serological, "blow-out", "to deliver," glass and plastic pipettes with a soft, precise, air-tight seal.
Catalog No. 50-145-7690 Supplier Thomas Scientific Supplier No. 1191K62
ProPette™ MPC is the precise and easy to use manual pipette controller that 's always ready to use. After attaching a pipette, one squeeze of its large rubber filling bulb is all it takes to start pipetting – there are no wires, no tubing and no battery to recharge! Begin aspirating by touching the EZ-Flow™ lever which is designed to precisely control the filling speed, based on how gently or firmly you press the lever. A built-in replaceable filter ensures optimum flow rate and protects the controller from liquid penetration and over-fill. A separate button provides a positive blow-out function when required. Made to accept pipettes from 0.1 to 100ml, the silicone "chuck" adapter is flexible, completely autoclavable, and will accommodate all types of serological, "blow-out", "to deliver," glass and plastic pipettes with a soft, precise, air-tight seal.